Orders LinkContactUsLinkComponent
Revised B/N surrogate mix
Revised B/N surrogate mix
Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 5 ml/ampule, 1
  REST31889.25
 :   Restek
Revised B/N surrogate mix
  REST31889.25
 :  Restek
 :  
Revised B/N surrogate mix
Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 5 ml/ampule, 1
 :  
1 * 1 Each
This mix contains four components (2-fluorobiphenyl, nitrobenzene-d5, p-terphenyl-d14, pyrene-d10).
  • Pakn:
    1
  • Beskrivelse:
    Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 5 ml/ampule