Revised B/N surrogate mix
Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 1 ml/ampule, 1
REST31888.25
: Restek
This mix contains four components (2-fluorobiphenyl, nitrobenzene-d5, p-terphenyl-d14, pyrene-d10).
- Pakn:1
- Beskrivelse:Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 1 ml/ampule