Orders LinkContactUsLinkComponent
Revised B/N surrogate mix
Revised B/N surrogate mix
Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 1 ml/ampule, 1
Katalog # REST31888.15
Leverandør:   Restek
Revised B/N surrogate mix
Katalog # REST31888.15
Leverandør:  Restek
CAS nummer:  
Revised B/N surrogate mix
Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 1 ml/ampule, 1
CAS nummer:  
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
This mix contains four components (2-fluorobiphenyl, nitrobenzene-d5, p-terphenyl-d14, pyrene-d10).
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Revised B/N Surrogate Mix, 5000 µg/ml, methylene chloride, 1 ml/ampule
Specifikationer