Sample bottles for ASE® 100/300 systems
250 ml, ravfarvet
Katalog # REST26260
Leverandør: Restek
Gå til:
Product Details & Documents
Sample collection bottles are made of glass. Caps and PTFE-lined septa included. Suitable for ASE 100/300 systems.
Specifikationer
- Kapacitet:250 ml
- Farve:ravfarvet