Orders LinkContactUsLinkComponent
Sample bottles for ASE® 100/300 systems
Sample bottles for ASE® 100/300 systems
250 ml, ravfarvet
Katalog # REST26260
Leverandør:   Restek
Sample bottles for ASE® 100/300 systems
Katalog # REST26260
Leverandør:  Restek
Sample bottles for ASE® 100/300 systems
Sample bottles for ASE® 100/300 systems
250 ml, ravfarvet
Price
426,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents
Sample collection bottles are made of glass. Caps and PTFE-lined septa included. Suitable for ASE 100/300 systems.
Specifikationer
  • Kapacitet:
    250 ml
  • Farve:
    ravfarvet