Resprep® tools for ASE® Systems
Filter insertion attachments for ASE 100/300 (use with 10 ml, 34 ml, 66 ml and 100 ml size cell bodies) and ASE 200 (use with 5 ml, 11 ml, 22 ml and 33 ml size cell bodies)
Katalog # REST26183
Leverandør: Restek
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Filter insertion attachments for ASE 100/300 (use with 10 ml, 34 ml, 66 ml and 100 ml size cell bodies) and ASE 200 (use with 5 ml, 11 ml, 22 ml and 33 ml size cell bodies)
- Seq:03