Orders LinkContactUsLinkComponent
Single - component oxygenates solutions
Single - component oxygenates solutions
tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
Katalog # REST30631
Leverandør:   Restek
Single - component oxygenates solutions
Katalog # REST30631
Leverandør:  Restek
CAS nummer:  
Single - component oxygenates solutions
Single - component oxygenates solutions
tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
CAS nummer:  
Price
kr344.00
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifikationer
Specifikationer
  • Pakn:
    1 Ampoul
  • Standard:
    US EPA 502.2, 8021, 8206B
  • Beskrivelse:
    tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanol
  • Pk Info:
    1 ml/ampoule
Specifikationer