Single - component oxygenates solutions
tert-Amyl ethyl ether (TAEE), with 2000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
Katalog # REST30617
Leverandør: Restek
Gå til:
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifikationer
Specifikationer
- Pakn:1 Ampoul
- Standard:US EPA 502.2, 8021, 8206B
- Beskrivelse:tert-Amyl ethyl ether (TAEE), with 2000 µg/ml in purge and trap grade methanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST30617
- Beskrivelse:tert-Amyl ethyl ether (TAEE)
- CAS-nr:919-94-8
- Formel:C₇H₁₆O



