Solid phase extraction cartridges, Resprep®, method specific
Ultra Quat SPE method, for use in HPLC analysis of paraquat/diquat, PP, 6 ml, 500 mg
Katalog # REST25499
Leverandør: Restek
Gå til:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifikationer
- Kapacitet:6 ml
- Beskrivelse:Ultra Quat SPE method, for use in HPLC analysis of paraquat/diquat
- Seq:01
- Lejevægt:500 mg
- Kolonnemateriale:PP