Valves for HPLC
Prime/purge valve repair kit for prime/purge valve, includes 2 soft seal, 2 PTFE O-ring, 7/32 hex key
Katalog # REST26407
Leverandør: Restek
Gå til:
Product Details & Documents
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
- Only fingertight torque is required to make the seal
Specifikationer
- Beskrivelse:Prime/purge valve repair kit for prime/purge valve, includes 2 soft seal, 2 PTFE O-ring, 7/32 hex key
- Seq:07