Orders LinkContactUsLinkComponent
Valves for HPLC
Prime/purge valve repair kit for prime/purge valve, includes 2 soft seal, 2 PTFE O-ring, 7/32 hex key
Katalog # REST26407
Leverandør:   Restek
Valves for HPLC
Katalog # REST26407
Leverandør:  Restek
Valves for HPLC
Valves for HPLC
Prime/purge valve repair kit for prime/purge valve, includes 2 soft seal, 2 PTFE O-ring, 7/32 hex key
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
  • Only fingertight torque is required to make the seal
Specifikationer
  • Beskrivelse:
    Prime/purge valve repair kit for prime/purge valve, includes 2 soft seal, 2 PTFE O-ring, 7/32 hex key
  • Seq:
    07