Warning signs
Warning hot surfaces, Polyester, 25×22 mm
Katalog # BRDY828830
Leverandør: Brady
Gå til:
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Specifikationer
- Beskrivelse:Warning hot surfaces
- Dimensioner:25×22 mm
- Materiale:Polyester
- Seq:04