Volumetric flasks, class A
50 ml, 12/14, 0,06 ± ml
Katalog # POULRB258/WAC/F/5
Leverandør: Poulten & Graf
Gå til:
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Specifikationer
- NS:12/14
- Kapacitet:50 ml
- Seq:07
- Tolerance:0,06 ± ml