Volumetric flasks, class A
10 ml, 10/13, 0,025 ± ml
Katalog # POULRB258/WAC/B/5
Leverandør: Poulten & Graf
Gå til:
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
POULRB258^WAC^J^2_TDS_EN
Datablad
Specifikationer
- NS:10/13
- Kapacitet:10 ml
- Seq:03
- Tolerance:0,025 ± ml