Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
5 ml, 10/13, 0,025 ± ml
Katalog # POULRB258/WAC/A/5
Leverandør:   Poulten & Graf
Volumetric flasks, class A
Katalog # POULRB258/WAC/A/5
Leverandør:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
5 ml, 10/13, 0,025 ± ml
Price
919,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifikationer
  • NS:
    10/13
  • Kapacitet:
    5 ml
  • Seq:
    02
  • Tolerance:
    0,025 ± ml