Volumetric flasks, class A
500 ml, 19/17, 0,25 ± ml
Catalog # POULRB258/J/2
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
POULRB258^WAC^J^2_TDS_EN
Datasheet
Specifications
- NS:19/17
- Kapacitet:500 ml
- Seq:27
- Tolerance:0,25 ± ml