Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
250 ml, 14/15, 0,15 ± ml
Katalog # POULRB258/I/2
Leverandør:   Poulten & Graf
Volumetric flasks, class A
Katalog # POULRB258/I/2
Leverandør:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
250 ml, 14/15, 0,15 ± ml
Price
546,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifikationer
  • NS:
    14/15
  • Kapacitet:
    250 ml
  • Seq:
    26
  • Tolerance:
    0,15 ± ml