Digital block heaters
Product Details & Documents
Versatile digital block heaters offer a wide range of blocks to accommodate various tubes provide temperature uniformity and stability for reproducible results. The units have digital controls and display of time and temperature and are durable with a powder-coated steel body. Ideal for sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation. Choice of models to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
- Built-in temperature sensing probe for outstanding temperature accuracy and control
- Precise temperature control with PID circuit
- Temperature calibration enables user to offset the temperature to desired value
- Timer allows the user to accurately monitor the heating time
- Interchangeable aluminum alloy sample blocks provide versatility and allow for easy cleaning and disinfecting
Leveringsinformation: Supplied with block extraction tool, but without blocks, these must be ordered separately.
Advarsel: * RT = Ambient
Note: Temp. stability value shown above is at 37 °C.
Specifikationer
- Opvarmnings kapacitet:400 W
- Antal blokke:2
- Temperaturområde:RT* +5...130 °C
- Vægt:3,3 kg
- Seq:02
- B×D×H:318×200×100 mm
- Temp. stabilitet:≤±1 °C
- Model:Block heater, digital
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- PT1000 temperature probe
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium blockAluminium block460-02990,5 ml + 0,2 ml Ø tubes30+20Log ind for tilgængelighed1 * 1 Items - 1.290,00 kr
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium blockAluminium block460-029825 mm + 12-13 mm + 6 mm Ø tubes3+12+6Log ind for tilgængelighed1 * 1 Items - 1.290,00 kr
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium blockAluminium block460-02971,5 ml + 2,0 ml tubes18+10Log ind for tilgængelighed1 * 1 Items - 940,00 kr
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Mixed aluminium blockMixed aluminium block460-02966 mm + 10 mm Ø tubes32+21Log ind for tilgængelighed1 * 1 Items - 940,00 kr
Produktinformation
- Katalog #TilB×D×HTilgængelighedPrice
- Aluminium blockAluminium block460-029596-well microplate124×76×39 mmLog ind for tilgængelighed1 * 1 Items - 664,00 kr
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium block
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium blockAluminium block460-03051×96 well PCR plate, half or full skirted-Log ind for tilgængelighed1 * 1 Items - 1.990,00 kr
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium blockAluminium block460-03041×96-well PCR plate, non skirted-Log ind for tilgængelighed1 * 1 Items - 1.740,00 kr
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium blockAluminium block460-030350 ml conical bottom tubes4Log ind for tilgængelighed1 * 1 Items - 3.120,00 kr
Produktinformation
- Katalog #TilAnt. hullerTilgængelighedPrice
- Aluminium blockAluminium block460-030215 ml conical bottom tubes15Log ind for tilgængelighed1 * 1 Items - 3.120,00 kr