Orders LinkContactUsLinkComponent
Carts
Carts
Starsys cart with interchangeable drawers, color-coded drawer pulls and Label holders available to quickly identify drawer contents, 578×632×1060 mm
Katalog # METOSXRSBED
Leverandør:   Metro International
Carts
Katalog # METOSXRSBED
Leverandør:  Metro International
Carts
Carts
Starsys cart with interchangeable drawers, color-coded drawer pulls and Label holders available to quickly identify drawer contents, 578×632×1060 mm
Price
62.530,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Starsys carts are available in a wide variety of heights and system widths and are ideal for the storage needs in the laboratories. The carts features smooth polymer top/worksurface and are provided with 127 mm polymer swivel casters, the front two casters comes with total-lock toe brake to lock both the wheel and the horn.
  • Easy to clean
  • Interchangeable drawers (76, 152 and 230 mm) with removable totes can be fully extended for easy access
  • Colour-coded drawer pulls are available in an array of choices
  • Label holders available to quickly identify drawer contents
Specifikationer
  • Beskrivelse:
    Starsys cart with interchangeable drawers, color-coded drawer pulls and Label holders available to quickly identify drawer contents
  • B×D×H:
    578×632×1060 mm