Accessories for Falcon® pipette controller
Hydrophobic filter, PTFE, 0,45 µm
Leverandør: Corning
Accessories for Falcon® pipette controller
Katalog # BDAA357473
Leverandør: Corning
Gå til:
Product Details & Documents
Documents
Specifikationer
- Beskrivelse:Hydrophobic filter, PTFE, 0,45 µm
- Seq:10
- Cat. No.:BDAA357473
- Miljøcertificering:I-RECs, ISO 14001 - Warsaw
- Environmentally Preferable:Lav fremstillingspåvirkning