Orders LinkContactUsLinkComponent
Multi-element standards, comprehensive mix B, VHG
Multi-element standards, comprehensive mix B, VHG
Comprehensive mix B standard, 1× 500 ml, 500 mL
Catalog # PROOVHG-SM90C-500
Multi-element standards, comprehensive mix B, VHG
Catalog # PROOVHG-SM90C-500
CAS Number:  
Multi-element standards, comprehensive mix B, VHG
Comprehensive mix B standard, 1× 500 ml, 500 mL
CAS Number:  
Price
14.260,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi element standards for Ag, Al, As, Ba, Bi, Ca, Cd, Ce, Dy, Er, Eu, Ga, Gd, Ho, La, Lu, Mg, Na, Nd, P, Pb, Pr, Rb, Sc, Se, Sm, Sr, Tb, Th, Tl, Tm, U, Y and Yb at 10 µg/ml in 40% aqua regia, tr. HF.
Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.
Specifications
Specifications
  • Pakn:
    500 mL
  • Beskrivelse:
    Comprehensive mix B standard
  • Pk Info:
    1× 500 ml