Spill kits, ADR
Large ADR maintenance spill kit, 63 litres, includes 35 pads (410×510 mm), 1 soc (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (250×250 mm), 1 pair gloves and 2 disposal bags
Katalog # 121-0153
Leverandør: Brady
Gå til:
Product Details & Documents
ADR spill kits are eco-friendly and portable and come in a durable see-through bag with shoulder strap. The small ADR chemical kit is designed for small spills of chemical and hazardous liquid. The large ADR maintenance kit is designed for small spills of industrial oil, water, solvents and coolants. The Re-Form ADR kit S is designed for small spills of industrial liquids except for highly aggressive acids and caustics.
- Quick and easy access to the sorbents
- Durable see-through bag makes it easy to check the spill response supplies
Specifikationer
- Beskrivelse:Large ADR maintenance spill kit, 63 litres, includes 35 pads (410×510 mm), 1 soc (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (250×250 mm), 1 pair gloves and 2 disposal bags
- Seq:03