Accessories: Light and control
Backlight stage, BL-CDW, with BF and DF techniques for transmitted-light microscopy, compatible with all Dino-Lite models (except long working distance models)
IDCPBL-CDW
- Beskrivelse:Backlight stage, BL-CDW, with BF and DF techniques for transmitted-light microscopy, compatible with all Dino-Lite models (except long working distance models)