Orders LinkContactUsLinkComponent
Watch glasses
Watch glass, PYREX® borosilicate glass, 80 mm
Katalog # BIBB1060/10D
Leverandør:   SciLabware
Watch glasses
Katalog # BIBB1060/10D
Leverandør:  SciLabware
Watch glasses
Watch glasses
Watch glass, PYREX® borosilicate glass, 80 mm
Price
1.270,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
  • Meget god kemisk resistens
  • Høj temperaturresistens
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifikationer
  • Ø:
    80 mm
  • Beskrivelse:
    Watch glass, PYREX® borosilicate glass
  • Seq:
    06