Watch glasses
Watch glass, PYREX® borosilicate glass, 60 mm
Catalog # BIBB1060/09D
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
- Meget god kemisk resistens
- Høj temperaturresistens
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifications
- Ø:60 mm
- Beskrivelse:Watch glass, PYREX® borosilicate glass
- Seq:04