Volumetric flasks, class A, white markings
1000 ml, 24/29, ± 0,40 ml
POULUS258/AM/WACK2
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
: Acc. to ISO and ASTM or USP standards
- NS:24/29
- Kapacitet:1000 ml
- Seq:35
- Tolerance:± 0,40 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



