Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Volumetric flasks, class A, white markings

250 ml, 14/23, ± 0,15 ml
Katalog # POULUS258/AM/WACI2
Leverandør:   Poulten & Graf
Volumetric flasks, class A, white markings
Katalog # POULUS258/AM/WACI2
Leverandør:  Poulten & Graf
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings

250 ml, 14/23, ± 0,15 ml

Price
1.030,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents

Borosilicate glass, amber, with PP NS-stopper.

  • Easy to read white markings
  • Very good chemical resistance
  • Very good temperature resistance

Amber coloured to protect light sensitive substances.

 : Acc. to ISO and ASTM or USP standards

Specifikationer
  • NS:
    14/23
  • Kapacitet:
    250 ml
  • Seq:
    33
  • Tolerance:
    ± 0,15 ml