Volumetric flasks, class A, white markings
200 ml, 14/23, ± 0,15 ml
Katalog # POULUS258/AM/WACH2
Leverandør: Poulten & Graf
Gå til:
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
: Acc. to ISO and ASTM or USP standards
Specifikationer
- NS:14/23
- Kapacitet:200 ml
- Seq:32
- Tolerance:± 0,15 ml



