Accessories
Cleaning tool (one tube holder, three tubes with caps) for use T 25 digital, T 25 easy clean digital/control homogenisers. Recommended cleaning process is: Pre-clean with suitable solvent, clean with detergent and final clean and rinse with demineralised water
Katalog # IKAA20003013
Leverandør:   IKA
Accessories
Katalog # IKAA20003013
Leverandør:  IKA
Accessories
Cleaning tool (one tube holder, three tubes with caps) for use T 25 digital, T 25 easy clean digital/control homogenisers. Recommended cleaning process is: Pre-clean with suitable solvent, clean with detergent and final clean and rinse with demineralised water
Price
1.010,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Cleaning tool (one tube holder, three tubes with caps) for use T 25 digital, T 25 easy clean digital/control homogenisers. Recommended cleaning process is: Pre-clean with suitable solvent, clean with detergent and final clean and rinse with demineralised water