Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
BUFFER SOLUTION PH 12.00 (20°C) 1 * 250 mL

BUFFER SOLUTION PH 12.00 (20°C) 1 * 250 mL

Katalog # APPC272586.1209
Leverandør:   PanReac AppliChem
BUFFER SOLUTION PH 12.00 (20°C) 1 * 250 mL
Katalog # APPC272586.1209
Leverandør:  PanReac AppliChem
BUFFER SOLUTION PH 12.00 (20°C) 1 * 250 mL
Price
481,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.