Temperature sensors
Temperature probe, PT100 with four wire and cable (1,8 m), +250…+500 °C, 4×50 mm
Katalog # WINKWFMP1500-XX015
Leverandør: WINKLER
Gå til:
Product Details & Documents
Specifikationer
- Område:+250…+500 °C
- Beskrivelse:Temperature probe, PT100 with four wire and cable (1,8 m)
- Seq:02
- Ø×L:4×50 mm