Orders LinkContactUsLinkComponent
Support for chairs, Allround
Support for chairs, Allround
Allround upper body support, red leather
Katalog # 139-0474
Leverandør:   Salli Systems
Support for chairs, Allround
Katalog # 139-0474
Leverandør:  Salli Systems
Support for chairs, Allround
Support for chairs, Allround
Allround upper body support, red leather
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This support is ideal for precision work in which the working positions change and versatile support is needed for the upper body.
  • Support can be changed from left-handed to right-handed and vice versa
  • Support is attached to the gas cylinder of the chair, so it can be used with all Salli chair models (SwingFit, Swing, MultiAdjuster, Twin, Classic)
  • Support arch length approx. 61 cm, width approx. 8,5 cm
  • Support upholstered with leather or artifical leather
Excellent for dentists, dental surgeons or dental assistants. It can also be used as a stretching support by moving it to the backside of the chair.
Specifikationer
  • Beskrivelse:
    Allround upper body support, red leather