Orders LinkContactUsLinkComponent
G-Biosciences Acetate Buffer
G-Biosciences Acetate Buffer
Acetate buffer, pH 5,5 (200 mM), 250 ml
Katalog # GENO786-877
Leverandør:   G-Biosciences
G-Biosciences Acetate Buffer
Katalog # GENO786-877
Leverandør:  G-Biosciences
CAS nummer:  
G-Biosciences Acetate Buffer
Acetate buffer, pH 5,5 (200 mM), 250 ml
CAS nummer:  
Price
532,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents
Acetate buffer at pH 5,0 and 5,5 are supplied as ready to use solutions that eliminate the calculation, formulation and weighing errors.
  • Precision in pH
  • Guaranteed reproducibility
Acetate buffer, pH 5,0 buffer is commonly used in protein purification, enzyme stabilisation, and histology.
Acetate buffer, pH 5,5 is useful for preserving enzyme activity or biomolecules that are sensitive to lower pH.
Specifikationer
Specifikationer
  • Beskrivelse:
    Acetate buffer, pH 5,5 (200 mM)
  • Volumen:
    250 ml
Specifikationer