X-Gal
Product Details & Documents
5-Bromo-4-chloro-3-indolyl beta-D-galactopyranoside is most commonly known as X-Gal. X-gal is a histochemical substrate for beta-galactosidase which cleaves X-Gal to yield an insoluble blue precipitate.
Molecular Biology Grade
- Purity: Greater than 99% (HPLC)
- Solution: Clear (2% in DMF)
- Biological Activity: Blue colour with Lambda, pUC, and M13
X-Gal is a mainstay in molecular biology cloning applications in which it is used to detect the activity of beta-galactosidase. X-Gal is used to detect the insertion of foreign DNA into the lacZ region of a plasmid DNA. Insertion of DNA into the lacZ region results in the loss of beta-galactosidase activity. Bacteria cells that retain active beta-galactosidase will result in characteristic blue colonies. Successful disruption of the lacZα gene disrupts the α-complemtation of the beta-galactosidase gene and the precipitate does not form, resulting in white colonies.
Many other applications also use X-Gal as a substrate to detect beta-galactosidase activity. These include beta-galactosidase -antibody linked immunoassays and immunohistochemistry, coliphage detection based on beta-galactosidase induction and the detection of micrometastasis formation during tumor progession.
X-gal is commonly used in conjunction with IPTG for blue/white screening.
- CAS nummer:7240-90-6
- Formel:C₁₄H₁₅BrClNO₆
- MDL nummer:MFCD00005666
- Molecular Weight:408,63 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:10 g



