Organic reference standards, sumatriptans, Ph. Eur., RS
Sumatriptan for system suitability, Reference standard, 2 mg
Katalog # EDQMY0000027
Leverandør: EDQM
Organic reference standards, sumatriptans, Ph. Eur., RS
Katalog # EDQMY0000027
Leverandør: EDQM
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakn:2 mg
- Standard:Reference standard
- Beskrivelse:Sumatriptan for system suitability
- Cat. No.:EDQMY0000027
- Beskrivelse:Sumatriptan for system suitability
- CAS-nr:103628-48-4
- Formel:C₁₈H₂₇N₃O₆S
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



