Orders LinkContactUsLinkComponent
Supelco® Spectroquant® Test Kits, Phosphate
Supelco® Spectroquant® Test Kits, Phosphate
Fosfatcelletest (PMB), 3,0 - 100 mg/L PO₄ - P, 9 - 307 mg/L PO₄, 7 - 229 mg/L P₂O₅, Phosphormolybdenum blue, Prove, Nova 30/60, Multy, Move, 25 Tests
Catalog # 1.00673.0001
Supplier:   Merck
Supelco® Spectroquant® Test Kits, Phosphate
Catalog # 1.00673.0001
Supplier:  Merck
CAS Number:  
Supelco® Spectroquant® Test Kits, Phosphate
Supelco® Spectroquant® Test Kits, Phosphate
Fosfatcelletest (PMB), 3,0 - 100 mg/L PO₄ - P, 9 - 307 mg/L PO₄, 7 - 229 mg/L P₂O₅, Phosphormolybdenum blue, Prove, Nova 30/60, Multy, Move, 25 Tests
CAS Number:  
Price
1.350,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 Denne metode er officielt anerkendt af USEPA som en alternativ metode til undersøgelse af følgende:
*7-1 spildevand
*7-2 drikkevand
*7-3 hhv. drikke- og spildevand
Specifications
Specifications
  • Kemisk metode:
    Phosphormolybdenum blue
  • Pakn:
    25 Tests
  • Beskrivelse:
    Fosfatcelletest (PMB)
  • Til enhed:
    Prove, Nova 30/60, Multy, Move
  • Område:
    3,0 - 100 mg/L PO₄ - P, 9 - 307 mg/L PO₄, 7 - 229 mg/L P₂O₅
Specifications