Dip slides
Plate count agar with TTC, neutraliser and VRBG agar, 20
Katalog # APPC435897.0922
Leverandør: PanReac AppliChem
Gå til:
Product Details & Documents
Dip slides are suitable for the microbiological hygiene control applications in both the food industry and other industrial applications.
- Easy to use
- Plate count ager for total aerobe count
- Rose bengal agar for moulds and yeasts count
- VRBG Agar for enterobacteria count
Each dip slide has two faces coated with agar-based medium, with a usable area of 12 cm² per face. Flexibility of the plastic strip allows samples to be taken in areas that are inaccessible to contact plates.
Supplied in a cardboard box and are protected by a stamped support and a sponge pad to absorb knocks and reduce the risk of the medium becoming detached.
Specifikationer
Specifikationer
- Pakn:20
- Beskrivelse:Plate count agar with TTC, neutraliser and VRBG agar