Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Plate count agar with TTC, neutraliser and Rose Bengal agar with chloramphenicol, 20
Katalog # APPC435896.0922
Leverandør:   PanReac AppliChem
Dip slides
Katalog # APPC435896.0922
Leverandør:  PanReac AppliChem
CAS nummer:  
Dip slides
Dip slides
Plate count agar with TTC, neutraliser and Rose Bengal agar with chloramphenicol, 20
CAS nummer:  
Price
1.020,00 kr
Log ind for tilgængelighed
1 * 20 Items
Product Details & Documents
Dip slides are suitable for the microbiological hygiene control applications in both the food industry and other industrial applications.
  • Easy to use
  • Plate count ager for total aerobe count
  • Rose bengal agar for moulds and yeasts count
  • VRBG Agar for enterobacteria count
Each dip slide has two faces coated with agar-based medium, with a usable area of 12 cm² per face. Flexibility of the plastic strip allows samples to be taken in areas that are inaccessible to contact plates.
Supplied in a cardboard box and are protected by a stamped support and a sponge pad to absorb knocks and reduce the risk of the medium becoming detached.
Specifikationer
Specifikationer
  • Pakn:
    20
  • Beskrivelse:
    Plate count agar with TTC, neutraliser and Rose Bengal agar with chloramphenicol
Specifikationer