Orders LinkContactUsLinkComponent
BUFFER SOLUTION PH 7.00 (20°C) (YELLOW C 1 * 1.000 mL
BUFFER SOLUTION PH 7.00 (20°C) (YELLOW C 1 * 1.000 mL
Katalog # APPC273617.1211
Leverandør:   PanReac AppliChem
BUFFER SOLUTION PH 7.00 (20°C) (YELLOW C 1 * 1.000 mL
Katalog # APPC273617.1211
Leverandør:  PanReac AppliChem
BUFFER SOLUTION PH 7.00 (20°C) (YELLOW C 1 * 1.000 mL
Price
522,00 kr
Log ind for tilgængelighed
1 * 1.000 mL
Product Details & Documents
Documents
Certifikater
Søg efter partinummer