Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for test sieves

Accessories for test sieves

Sieve pan with discharge nozzle, 300 mm, 60 mm
Catalog # HAVE205922855
Supplier:   NEXOPART
Accessories for test sieves
Catalog # HAVE205922855
Supplier:  NEXOPART
Accessories for test sieves
Accessories for test sieves

Sieve pan with discharge nozzle, 300 mm, 60 mm

Price
2.180,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Ø:
    300 mm
  • Beskrivelse:
    Sieve pan with discharge nozzle
  • Højde:
    60 mm
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.