Venepunktursæt, Surshield™ Surflo®
Winged infusion sets with protector, G 23, lyseblå, 19 mm
Katalog # TERUSV-S23NL30
Leverandør: TERUMO
Gå til:
Product Details & Documents
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.
Specifikationer
- Måler:G 23
- Farve:lyseblå
- Beskrivelse:Winged infusion sets with protector
- Længde:19 mm
- Seq:003