Skip to Header
Skip to Main Content
Skip to Footer
Udvidet søgning
SDS
Certificates
Producentdata
0
0,00 kr
Ordreindtastning
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Hjem
BUFFER PH 4,01 1 * 1.000 mL
Print
BUFFER PH 4,01 1 * 1.000 mL
Catalog # KNICCS-P0400/1000
Supplier:
KNICK
BUFFER PH 4,01 1 * 1.000 mL
Catalog # KNICCS-P0400/1000
Supplier:
KNICK
Return to Top
BUFFER PH 4,01 1 * 1.000 mL
Price
434,00 kr
Log in
for availability
1 * 1.000 mL
-
+
Return Policy
Product Details & Documents