pH-elektroder
SE 106 N, combination pH/PT1000 electrode, SE 106 N, 0...14, −5...+100 °C, Glas, 3 mol/l KCl, Ground glass sleeve, 12×170 mm, DIN, Difficult samples, low conductivity samples, coating media
Katalog # KNICSE106N
Leverandør: KNICK
Gå til:
Product Details & Documents
Specifikationer
- Tilslutning:DIN
- Ø×L:12×170 mm
- Til:Difficult samples, low conductivity samples, coating media
- Model:SE 106 N
- Beskrivelse:SE 106 N, combination pH/PT1000 electrode
- Materiale:Glas
- Seq:07
- Membran:Ground glass sleeve
- Elektrolyt:3 mol/l KCl
- Temperatur:−5...+100 °C
- pH:0...14