Accessories for test sieves
Sieve pan with discharge nozzle, 200 mm, 50 mm
Katalog # HAVE205922824
Leverandør: NEXOPART
Gå til:
Product Details & Documents
Specifikationer
- Ø:200 mm
- Beskrivelse:Sieve pan with discharge nozzle
- Højde:50 mm
- Seq:01