High visibility shorts, E043
fluorescerende gul/mørkeblå, S
Katalog # PRTWE043YGYS
Leverandør: Portwest
Gå til:
Product Details & Documents
Knee-length summer shorts made of durable polyester/cotton mix, with Texpel stain-resistant finish.
- Two side pockets, two back patch pockets
- Half elasticated waistband
- Thigh pocket, velcro fastened with mobile phone pocket
- Rule pocket
Weight: 245 g/m²
Certifications: EN ISO 20471 (Class 1)
Specifikationer
- Tøj størrelse:S
- Farve:fluorescerende gul/mørkeblå
- Seq:06
- Køn:Unisex
- Cat. No.:PRTWE043YGYS
- Sleeve or leg length:Short
- Risk area:HiVis
- Vægt:245 g/m²
- Model:E043
- Outfit applikationer:General Purpose
- Stof:Polyester/cotton