Signs, warning stands
Rutschgefahr (german), PP, -
BRDY241930
These stable warning devices are easy to transport and suitable for the marking of hazardous zones.
- Made of high-density polypropylene
- Durable, yellow
- No buckling or corrosion
- Large note field is visible from over 9 meters
- With default notices or unlabeled also available
Height: 660 mm
- Beskrivelse:Rutschgefahr (german)
- Dimensioner:-
- Materiale:PP
- Seq:03