Orders LinkContactUsLinkComponent
VWR® VisiScope®, CCD Camera Adapters
Ring adapter for mono- and binocular microscopes (Ø = 30 mm)
  630-2412
VWR® VisiScope®, CCD Camera Adapters
  630-2412
VWR® VisiScope®, CCD Camera Adapters
VWR® VisiScope®, CCD Camera Adapters
Ring adapter for mono- and binocular microscopes (Ø = 30 mm)
56,00 kr
1 * 1 Items
  • Beskrivelse:
    Ring adapter for mono- and binocular microscopes (Ø = 30 mm)
  • Seq:
    300