Orders LinkContactUsLinkComponent
Phosphate-buffered saline (PBS), Corning®
Phosphate-buffered saline (PBS), Corning®
PBS (Phosphate-Buffered Saline) 1X without calcium or magnesium, 6×500 ml, 6
Katalog # 21-040-CV
Leverandør:   Corning
Phosphate-buffered saline (PBS), Corning®
Katalog # 21-040-CV
Leverandør:  Corning
CAS nummer:  
Phosphate-buffered saline (PBS), Corning®
Phosphate-buffered saline (PBS), Corning®
PBS (Phosphate-Buffered Saline) 1X without calcium or magnesium, 6×500 ml, 6
CAS nummer:  
Price
435,00 kr
Log in for availability
1 * 6 Items
Product Details & Documents
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate
  • Low endotoxin - <0.25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
PBS helps to maintain a constant pH and is isotonic and non-toxic to cells.
Specifications
Specifications
  • Pakn:
    6
  • Beskrivelse:
    PBS (Phosphate-Buffered Saline) 1X without calcium or magnesium, 6×500 ml