Orders LinkContactUsLinkComponent
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), powder, with Earle's salts and L-glutamine without sodium bicarbonate, Powder, 10 L
Katalog # 50-050-PB
Leverandør:   Corning
Medium 199 (Mod.), Corning®
Katalog # 50-050-PB
Leverandør:  Corning
CAS nummer:  
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), powder, with Earle's salts and L-glutamine without sodium bicarbonate, Powder, 10 L
CAS nummer:  
Price
401,00 kr
Log ind for tilgængelighed
1 * 10 L
Product Details & Documents
Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.
Specifikationer
Specifikationer
  • Medieformat:
    Powder
  • Pakn:
    10 L
  • Beskrivelse:
    Medium 199 (Mod.), powder, with Earle's salts and L-glutamine without sodium bicarbonate