Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sodium bicarbonate, Corning®

Sodium bicarbonate, Corning®

Sodium bicarbonate, powder, 500 g
Katalog # MDTC61-065-RO
Leverandør:   Corning
Sodium bicarbonate, Corning®
Katalog # MDTC61-065-RO
Leverandør:  Corning
CAS nummer:  
Sodium bicarbonate, Corning®
Sodium bicarbonate, Corning®

Sodium bicarbonate, powder, 500 g

CAS nummer:  
Price
734,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Sodium bicarbonate is used as part of a buffering system commonly used to maintain physiological pH 7,2 to 7,4 in a culture environment

Bicarbonate is a weak buffer with that makes maintaining a pH range of 7,2 to 7,6 somewhat difficult, but it is non-toxic, and is a nutrient source.

Specifikationer

Specifikationer

  • Pakn:
    500 g
  • Beskrivelse:
    Sodium bicarbonate, powder
  • Cat. No.:
    MDTC61-065-RO
  • Opbevaringstemperatur:
    Ambient

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.