Organic reference standards, Formaldehyde IC Standards, Supelco®
Formaldehyde IC Standard, 1000 μg/ml in H₂O, Analytical standard, 500 mL
Katalog # ICS045-500ML
Leverandør: Merck
Organic reference standards, Formaldehyde IC Standards, Supelco®
Katalog # ICS045-500ML
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Koncentration:1000 μg/ml in H₂O
- Pakn:500 mL
- Standard:Analytical standard
- Beskrivelse:Formaldehyde IC Standard
- Cat. No.:ICS045-500ML
- Beskrivelse:Formaldehyde IC Standard
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



