KLEEN blade management system
The KLEEN Blade Management System allows for effortless safe attachment and removal of surgical blades from a standard handle. The system which incorporates a quality Swann-Morton surgical blade is designed to operate with most No.3 and No.4 standard surgical handles already in circulation within the hospital system thus minimising disruption and cost during the conversion process.
- Sterile and non sterile blades
- Protective sliding sheath
The sheath covers the blade completely thus assisting with accident free handling. It fully exposes the surgical blade when in use with an intuitive lock which prevents any forward movement of the sheath during the procedure. Easy to detach the BMS encapsulates the contaminated sharp which is still visible for the final count and to help ensure safe and responsible disposal.
- Beskrivelse:Kleen BMS No. 11
- Materiale:Rustfrit stål
- Seq:02